Skip to Content
Merck
All Photos(1)

Key Documents

Y0000116

Acamprosate impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3-Amino-1-propanesulfonic acid, 3APS, Homotaurine, Tramiprosate

Sign Into View Organizational & Contract Pricing

Select a Size

110 MG
$218.00

$218.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
110 MG
$218.00

About This Item

Linear Formula:
H2N(CH2)3SO3H
CAS Number:
Molecular Weight:
139.17
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$218.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

acamprosate

manufacturer/tradename

EDQM

mp

293 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NCCCS(O)(=O)=O

InChI

1S/C3H9NO3S/c4-2-1-3-8(5,6)7/h1-4H2,(H,5,6,7)

InChI key

SNKZJIOFVMKAOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Acamprosate impurity A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gunnar K Gouras et al.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics, 12(1), 3-11 (2014-11-06)
Many lines of evidence support that β-amyloid (Aβ) peptides play an important role in Alzheimer's disease (AD), the most common cause of dementia. But despite much effort the molecular mechanisms of how Aβ contributes to AD remain unclear. While Aβ
Karin Ortmayr et al.
Nature communications, 10(1), 1841-1841 (2019-04-25)
Transcriptional reprogramming of cellular metabolism is a hallmark of cancer. However, systematic approaches to study the role of transcriptional regulators (TRs) in mediating cancer metabolic rewiring are missing. Here, we chart a genome-scale map of TR-metabolite associations in human cells
Shuangchan Wu et al.
Neuropharmacology, 83, 107-117 (2014-04-29)
Tramiprosate, a small aminosulphonate compound, is present in various species of red marine algae. In this study, we examined whether tramiprosate protects neurons and improves functional recovery following ischemic stroke in rats subjected to the intraluminal filament model of MCAO
Pieter Glibert et al.
Journal of proteome research, 14(2), 839-849 (2014-12-17)
The ability to distinguish between phosphopeptides of high and low stoichiometry is essential to discover the true extent of protein phosphorylation. We here extend the strategy whereby a peptide sample is briefly split in two identical parts and differentially labeled

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service