Skip to Content
Merck
All Photos(1)

Key Documents

Y0000110

Adipic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Hexanedioic acid

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$218.00

$218.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$218.00

About This Item

Linear Formula:
HOOC(CH2)4COOH
CAS Number:
Molecular Weight:
146.14
Beilstein:
1209788
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$218.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

Agency

EP Reference Standard

vapor density

5 (vs air)

vapor pressure

1 mmHg ( 159.5 °C)

API family

adipic acid

autoignition temp.

788 °F

manufacturer/tradename

EDQM

bp

265 °C/100 mmHg (lit.)

mp

151-154 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)CCCCC(O)=O

InChI

1S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)

InChI key

WNLRTRBMVRJNCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Adipic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

384.8 °F - closed cup

Flash Point(C)

196 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tino Polen et al.
Journal of biotechnology, 167(2), 75-84 (2012-07-25)
Adipic acid is the most important commercial aliphatic dicarboxylic acid in the chemical industry and is primarily used for the production of nylon-6,6 polyamide. The current adipic acid market volume is about 2.6 million tons/y and the average annual demand
Kuo Chu Hwang et al.
Science (New York, N.Y.), 346(6216), 1495-1498 (2014-12-20)
Nitric acid oxidation of cyclohexane accounts for ~95% of the worldwide adipic acid production and is also responsible for ~5 to 8% of the annual worldwide anthropogenic emission of the ozone-depleting greenhouse gas nitrous oxide (N2O). Here we report a
Joseph A Majdi et al.
Investigative ophthalmology & visual science, 56(1), 587-597 (2014-12-20)
We developed a novel technique for accelerated drug screening and retinotoxin characterization using time-lapse optical coherence tomography (OCT) and a drug microapplication device. Using an ex vivo rabbit eyecup preparation, we studied retinotoxin effects in real-time by microperfusing small retinal
Mohamad F Aslam et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(8), 3671-3678 (2014-04-30)
The ferritin core is composed of fine nanoparticulate Fe(3+) oxohydroxide, and we have developed a synthetic mimetic, nanoparticulate Fe(3+) polyoxohydroxide (nanoFe(3+)). The aim of this study was to determine how dietary iron derived in this fashion is absorbed in the
Hong-Fei Wang et al.
Antonie van Leeuwenhoek, 107(3), 835-843 (2015-01-09)
A white bacterial strain, designated EGI 650022(T), was isolated from the roots of Salsola affinis C. A. Mey, collected from Urumqi City, Xinjiang, north-western China. The strain was found to be aerobic, Gram-stain positive, oxidase-positive and catalase-positive. Cells were non-motile

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service