Skip to Content
Merck
All Photos(1)

Key Documents

Y0000033

Carbamazepine impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

10,11-Dihydrocarbamazepine, 10,11-Dihydro-5H-dibenz[b,f]azepine-5-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
$218.00

$218.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 MG
$218.00

About This Item

Empirical Formula (Hill Notation):
C15H14N2O
CAS Number:
Molecular Weight:
238.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$218.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

carbamazepine

manufacturer/tradename

EDQM

mp

205-210 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(=O)N1c2ccccc2CCc3ccccc13

InChI

1S/C15H14N2O/c16-15(18)17-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)17/h1-8H,9-10H2,(H2,16,18)

InChI key

PHNLCHMJDSSPDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Carbamazepine impurity A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1A - Skin Sens. 1A - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M G Bixel et al.
European journal of biochemistry, 267(1), 110-120 (1999-12-22)
Several wasp venoms contain philanthotoxins (PhTXs), natural polyamine amides, which act as noncompetitive inhibitors (NCIs) on the nicotinic acetylcholine receptor (nAChR). Effects of varying the structure of PhTXs and poly(methylene tetramine)s on the binding affinity have been investigated. Using the
William T A Harrison et al.
Acta crystallographica. Section C, Crystal structure communications, 62(Pt 5), o240-o242 (2006-05-09)
The title compound (systematic name: 10,11-dihydro-5H-dibenz[b,f]azepine-5-carboxamide), C15H14N2O, is shown to crystallize as an orthorhombic polymorph to complement the known monoclinic form. The molecular conformations of both forms are very similar, involving a bent conformation for the seven-membered azepine ring and
T D Cyr et al.
Journal - Association of Official Analytical Chemists, 70(5), 836-840 (1987-09-01)
Liquid chromatographic (LC) methods have been developed for the determination of carbamazepine, the impurity 10,11-dihydrocarbamazepine, and related compounds in carbamazepine drug substance and tablets. The LC methods specify a 5 micron diol column and a mobile phase of acetonitrile-methanol-0.05% aqueous

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service