Recommended Products
description
non-ionic
technique(s)
ELISA: suitable
cell culture | mammalian: suitable
enzyme immunoassay: suitable
hybridization: suitable
protein purification: suitable
InChI
1S/C18H30O3/c1-17(2,3)14-18(4,5)15-6-8-16(9-7-15)21-13-12-20-11-10-19/h6-9,19H,10-14H2,1-5H3
InChI key
LBCZOTMMGHGTPH-UHFFFAOYSA-N
Legal Information
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
483.8 °F - closed cup
Flash Point(C)
251 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jairus Kleinert et al.
Biomicrofluidics, 9(3), 034104-034104 (2015-06-06)
The operation of digital microfluidic devices with water droplets manipulated by electrowetting is critically dependent on the static and dynamic stability and lubrication properties of the oil films that separate the droplets from the solid surfaces. The factors determining the
Abdullah Taner Bi Şgi N
Journal of AOAC International, 102(5), 1516-1522 (2019-05-16)
Background: Copper (Cu) is an essential metal for humans at certain concentrations. However, it can be toxic at higher concentrations. Therefore, determination of Cu content of foodstuff is important. Objective: The aim of the study was to develop a simple
Russell M Morphew et al.
Veterinary parasitology, 191(3-4), 379-385 (2012-10-02)
Fasciola hepatica NP-40 released protein extract (FhNPE) exhibits potent Th1 immunosuppressive properties in vitro and in vivo. However, the protein composition of this active fraction, responsible for Th1 immune modulatory activity, has yet to be resolved. Therefore, FhNPE, a Nonidet
Rui Ma et al.
Analytical biochemistry, 424(2), 124-126 (2012-03-01)
Triton X-114 temperature transition extraction has been considered to be a simple and cost-effective strategy to eliminate endotoxin from plasmid preparations. However, a repeated cooling-heating process may promote the degradation of plasmid DNA. Based on the finding that the cloud
Zonghao Li et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 67(2), 247-253 (2012-11-22)
A new, simple and rapid method based on dispersive liquid-liquid microextraction (DLLME) was developed for extracting and preconcentrating copper (Cu), nickel (Ni), lead (Pb) and cadmium (Cd) in water samples prior to flame atomic absorption spectrometry (FAAS) analysis. 1-(2-thiazolylazo)-naphthol (TAN)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service