Skip to Content
Merck
All Photos(1)

Key Documents

W429540

Sigma-Aldrich

Citronellyl tiglate

≥95%, stabilized, FG

Sign Into View Organizational & Contract Pricing

Select a Size

W429540-SAMPLE-K
$75.30
100 G
$307.00
1 KG
$1,970.00

$75.30


Estimated to ship on11 April 2025


Request a Bulk Order

Select a Size

Change View
W429540-SAMPLE-K
$75.30
100 G
$307.00
1 KG
$1,970.00

About This Item

Empirical Formula (Hill Notation):
C15H26O2
CAS Number:
Molecular Weight:
238.37
FEMA Number:
4295
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.340
NACRES:
NA.21
Organoleptic:
geranium; leafy; fruity; floral; rose
grade:
FG
Halal
Kosher
Agency:
follows IFRA guidelines
food allergen:
no known allergens

$75.30


Estimated to ship on11 April 2025


Request a Bulk Order

grade

FG
Halal
Kosher

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

contains

synthetic α-tocopherol as stabilizer

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

geranium; leafy; fruity; floral; rose

SMILES string

O(CCC(CCC=C(C)C)C)C(=O)\C(=C\C)\C

InChI

1S/C15H26O2/c1-6-14(5)15(16)17-11-10-13(4)9-7-8-12(2)3/h6,8,13H,7,9-11H2,1-5H3/b14-6+

InChI key

UCFQYMKLDPWFHZ-MKMNVTDBSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service