Skip to Content
Merck
All Photos(1)

Key Documents

W426060

Sigma-Aldrich

S-Isopropyl-3-methylbut-2-enethioate

≥95%, FG

Sign Into View Organizational & Contract Pricing

Select a Size

W426060-SAMPLE
$75.30
25 G
$150.00
100 G
$452.00

$75.30


Estimated to ship on10 May 2025


Request a Bulk Order

Select a Size

Change View
W426060-SAMPLE
$75.30
25 G
$150.00
100 G
$452.00

About This Item

Empirical Formula (Hill Notation):
C8H14OS
CAS Number:
Molecular Weight:
158.26
FEMA Number:
4260
MDL number:
UNSPSC Code:
12164502
Flavis number:
12.134
NACRES:
NA.21
Organoleptic:
green; alliaceous
grade:
FG
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

$75.30


Estimated to ship on10 May 2025


Request a Bulk Order

grade

FG

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; alliaceous

SMILES string

S(C(C)C)C(=O)C=C(C)C

InChI

1S/C8H14OS/c1-6(2)5-8(9)10-7(3)4/h5,7H,1-4H3

InChI key

IZVXTRDVESJRTA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service