Skip to Content
Merck
All Photos(1)

Key Documents

W0100000

Warfarin sodium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3-(α-Acetonylbenzyl)-4-hydroxycoumarin sodium salt, 4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$254.00

$254.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$254.00

About This Item

Linear Formula:
C19H15O4Na
CAS Number:
Molecular Weight:
330.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$254.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

warfarin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na].CC(=O)CC(c1ccccc1)C2=C(O)c3ccccc3OC2=O

InChI

1S/C19H16O4.Na/c1-12(20)11-15(13-7-3-2-4-8-13)17-18(21)14-9-5-6-10-16(14)23-19(17)22;/h2-10,15,21H,11H2,1H3;/q;+1/p-1

InChI key

KYITYFHKDODNCQ-UHFFFAOYSA-M

Gene Information

human ... VKORC1(79001)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Warfarin sodium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 3 - Repr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frieder Schlunk et al.
Translational stroke research, 6(2), 133-139 (2014-11-27)
Little is known about the pathophysiology of oral anticoagulation-associated intracerebral hemorrhage (OAC-ICH). We compared hematoma volume, number of terminal deoxynucleotidyl dUTP nick-end labeling (TUNEL)-positive cells (indicating cell death), MMP-9 levels, and perilesional edema formation between warfarin-treated mice and controls. Intracerebral
Catherine K Chang et al.
Journal of vascular surgery, 60(6), 1554-1564 (2014-07-22)
Despite poor long-term patency, acceptable limb salvage has been reported with cryopreserved saphenous vein bypass (CVB) for various indications. However, utility of CVB in patients with critical limb ischemia (CLI) remains undefined. The purpose of this analysis was to determine
Ryohei Hamaguchi et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 967, 110-117 (2014-08-03)
We developed a high-performance liquid chromatography-tandem mass spectrometric method for phospholipid biomarker discovery and applied it to a cell-based assay system for the screening of phospholipidosis-inducing drugs. We studied the compositions of phospholipid molecules exceeding 100 species in cultured cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service