Skip to Content
Merck
All Photos(5)

Key Documents

VT000302

Millipore

Salmonella enterica subsp. enterica serovar Enteritidis WDCM 00030 Vitroids

15-80 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

S. enterica WDCM 00030 Vitroids, Salmonella enterica subsp. enterica serovar Enteritidis WDCM 00030 Vitroids, Certified reference material, Microbiological standards

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$157.00

$157.00


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
10 EA
$157.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$157.00


Check Cart for Availability

Request a Bulk OrderRequest more information

grade

certified reference material

Quality Level

Agency

ISO 17034
ISO/IEC 17025

form

solid

shelf life

limited shelf life, expiry date on the label

packaging

pack of 10 discs

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

application(s)

microbiology

storage temp.

−20°C

suitability

Salmonella enterica
Salmonella spp.

Looking for similar products? Visit Product Comparison Guide

General description

Salmonella enterica subsp. enterica serovar Enteritidis WDCM 00030 Vitroids are Certified Microorganisms Standards (Certified Microbiological Reference Materials) which can be used as quantitative and qualitative control organisms. They contain colony forming units (CFUs) in the range of 15-80 CFU per disc. The technology allows long-term conservation and renders the organisms resistant to temperature changes. They are intended to be used as daily internal quality control organisms. These microorganisms are certified according to ISO/IEC 17025 and produced under reproducible conditions compliant with ISO 17034 using authenticated strains from CECT®. These ready-to-use microbiological controls minimize the need to maintain control strains in the test laboratory and guarantee an authenticated control culture for every quality control test. Vitroids are derived from a traceable culture obtained freeze-dried from CECT®.

Application

Salmonella enterica subsp. enterica serovar Enteritidis WDCM 00030 Vitroids are used as reference strains in routine quality control for food, beverage, water, and environmental testing. Additionally, they are excellent for use in method development and validations, as starter cultures, and for research studies and educational purposes.

Features and Benefits

  • Ready-to-use at convenient CFU ranges
  • No dilution required
  • Quick rehydration in 10 minutes
  • 2-3 years of shelf life in a standard -20 °C freezer
  • Complete with a comprehensive certificate of analysis
  • No maintenance of stock cultures
  • No recovery time or pre-enrichment step is necessary
  • Robust, efficient, and reliable
  • Easy-to-use
  • Cost-effective

Other Notes

Strain History

< CIP, 1986 < ATCC < CDC
Strain origin: CECT® 4300

Other cell culture collections:
  • DSM 9898
  • ATCC® 13076
Click here to access the BacDive database for more info on metabolism, genome, growth media, and more

Legal Information

Derived from CECT® strains
ATCC is a registered trademark of American Type Culture Collection
CECT is a registered trademark of Universitat de Valencia
Vitroids is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Explore certified microbiological reference materials: VITROIDS™, LENTICULE® discs by Millipore®. Ideal for food, beverage, cannabis testing. Order today.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service