Skip to Content
Merck
All Photos(2)

Key Documents

T6062

Sigma-Aldrich

Trifluoperazine hydrochloride

meets USP testing specifications

Synonym(s):

Trifluoperazine dihydrochloride, 10-[3-(4-Methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)-10H-phenothiazine dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$135.00

$135.00


Available to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
$135.00

About This Item

Empirical Formula (Hill Notation):
C21H24F3N3S · 2HCl
CAS Number:
Molecular Weight:
480.42
Beilstein:
3820024
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.21

$135.00


Available to ship on03 April 2025Details


Request a Bulk Order

Agency

USP/NF
meets USP testing specifications

Quality Level

Assay

98-101% dry basis

form

powder

mp

243 °C (dec.) (lit.)

solubility

water: 50 g/L, clear

application(s)

pharmaceutical (small molecule)

storage temp.

−20°C

SMILES string

Cl[H].Cl[H].CN1CCN(CCCN2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1

InChI

1S/C21H24F3N3S.2ClH/c1-25-11-13-26(14-12-25)9-4-10-27-17-5-2-3-6-19(17)28-20-8-7-16(15-18(20)27)21(22,23)24;;/h2-3,5-8,15H,4,9-14H2,1H3;2*1H

InChI key

BXDAOUXDMHXPDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Muta. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Antonia P Gunesch et al.
Antimicrobial agents and chemotherapy, 64(8) (2020-06-10)
Several cationic amphiphilic drugs (CADs) have been found to inhibit cell entry of filoviruses and other enveloped viruses. Structurally unrelated CADs may have antiviral activity, yet the underlying common mechanism and structure-activity relationship are incompletely understood. We aimed to understand
Stephanie N Brosius et al.
Journal of neuropathology and experimental neurology, 73(11), 1078-1090 (2014-10-08)
Chemotherapeutic agents effective against malignant peripheral nerve sheath tumors (MPNSTs) are urgently needed. We recently found that tamoxifen potently impedes xenograft growth. In vitro, tamoxifen inhibits MPNST proliferation and survival in an estrogen receptor-independent manner; these effects are phenocopied by
Ashleigh Pulkoski-Gross et al.
Molecular pharmacology, 87(3), 501-512 (2015-01-02)
Because cancer cell invasion is a critical determinant of metastasis, targeting invasion is a viable approach to prevent metastasis. Utilizing a novel three-dimensional high-throughput invasion assay, we screened a National Cancer Institute compound library and discovered compounds demonstrating inhibitory effects
Seung Jun Lee et al.
Toxicology letters, 232(2), 458-465 (2014-12-03)
In the present study, we evaluated the inhibitory potentials of finasteride for the major human hepatic UDP-glucuronosyltransferases (UGTs) (UGT1A1, UGT1A3, UGT1A4, UGT1A6, UGT1A9, UGT2B7, and UGT2B15) in vitro using LC-MS/MS by specific marker reactions in human liver microsomes (except for
Eu Jin Choi et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 72, 13-19 (2014-07-06)
In this study, we evaluated inhibitory potentials of popularly-consumed berries (bilberry, blueberry, cranberry, elderberry, and raspberry ketones) as herbal supplements on UGT1A1, UGT1A4, UGT1A6, UGT1A9, and UGT2B7 in vitro. We also investigated the potential herb-drug interaction via UGT1A1 inhibition by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service