Skip to Content
Merck
All Photos(1)

Key Documents

T2005010

Triflusal impurity B

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Hydroxy-4-(trifluoromethyl)benzoic acid, 4-(Trifluoromethyl)salicylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

15 MG
$204.00

$204.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
15 MG
$204.00

About This Item

Empirical Formula (Hill Notation):
C8H5F3O3
CAS Number:
Molecular Weight:
206.12
UNSPSC Code:
41116107
NACRES:
NA.24

$204.00


Estimated to ship on02 June 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

triflusal

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(c(cc1)C(=O)O)O

InChI

1S/C8H5F3O3/c9-8(10,11)4-1-2-5(7(13)14)6(12)3-4/h1-3,12H,(H,13,14)

InChI key

XMLFPUBZFSJWCN-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Triflusal impurity B EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Velasco et al.
The Journal of urology, 166(5), 1962-1968 (2001-10-05)
We examined the effects of intravenous administration of the 2 nuclear factor-kappaB inhibitors aspirin and 2-hydroxy-4-trifluoromethylbenzoic acid (HTB) on bladder filling and voiding in anesthetized and conscious rats. Disappearance of isovolumic bladder contractions after intravenous administration of different doses of
A Fernández de Arriba et al.
Molecular pharmacology, 55(4), 753-760 (1999-04-01)
The therapeutic potential of drugs that block the induction of cyclooxygenase-2 has been emphasized. When two 4-trifluoromethyl salicylate derivatives [2-acetoxy-4-trifluoromethyl-benzoic acid (triflusal) and its deacetylated metabolite 2-hydroxy-4-trifluoromethylbenzoic acid (HTB)] were compared with aspirin and sodium salicylate as cyclooxygenase-2 (COX-2) inhibitors
R Mis et al.
European journal of clinical pharmacology, 42(2), 175-179 (1992-01-01)
2-hydroxy-4-trifluoromethylbenzoic acid (HTB) is the main active metabolite of the platelet antiaggregant drug triflusal. Its binding to plasma proteins of rats and healthy volunteers in vitro and in vivo has been studied. Rats were given a single oral dose of
F Boscá et al.
Photochemistry and photobiology, 73(5), 463-468 (2001-05-23)
Triflusal is a platelet antiaggregant drug with photoallergic side effects. However, it is considered a prodrug since it is metabolized to 2-hydroxy-4-trifluoromethylbenzoic acid (HTB)--the pharmacologically active form. HTB was found to be photolabile under various conditions. Its major photodegradation pathway
J Ramis et al.
European journal of drug metabolism and pharmacokinetics, 16(4), 269-273 (1991-10-01)
The pharmacokinetic profile of triflusal (2-acetoxy-4-trifluoromethyl benzoic acid) and its main metabolite HTB (2-hydroxy-4-trifluoromethyl benzoic acid) has been studied in 8 healthy subjects (4 males and 4 females), after a single oral dose of 900 mg of triflusal. Plasma concentrations

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service