Skip to Content
Merck
All Photos(1)

Key Documents

T1977990

Tricaprin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Glyceryl tridecanoate, 1,2,3-Tricaprinoylglycerol, 1,2,3-Tridecanoylglycerol, Glycerol tricaprate, Glycerol tris(decanoate), Tricaprin, Tridecanoin

Sign Into View Organizational & Contract Pricing

Select a Size

310 MG
$265.00

$265.00


Available to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
310 MG
$265.00

About This Item

Linear Formula:
[CH3(CH2)8COOCH2]2CHOCO(CH2)8CH3
CAS Number:
Molecular Weight:
554.84
Beilstein:
1717683
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$265.00


Available to ship on17 April 2025Details


Request a Bulk Order

grade

pharmaceutical primary standard

API family

tricaprin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCC)OC(=O)CCCCCCCCC

InChI

1S/C33H62O6/c1-4-7-10-13-16-19-22-25-31(34)37-28-30(39-33(36)27-24-21-18-15-12-9-6-3)29-38-32(35)26-23-20-17-14-11-8-5-2/h30H,4-29H2,1-3H3

InChI key

LADGBHLMCUINGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tricaprin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Liu et al.
PloS one, 8(10), e77856-e77856 (2013-11-10)
Esterases with excellent merits suitable for commercial use in ester production field are still insufficient. The aim of this research is to advance our understanding by seeking for more unusual esterases and revealing their characterizations for ester synthesis. A novel
Pipsa Kulovesi et al.
Investigative ophthalmology & visual science, 55(7), 4448-4454 (2014-05-31)
The tear film lipid layer is believed to stabilize the tear film and to retard evaporation. Based on previous simple in vitro studies, the evidence for the latter property is scarce. In this study, we used complex lipid mixtures including

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service