Skip to Content
Merck
All Photos(1)

Key Documents

T1820000

Trapidil

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$204.00

$204.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$204.00

About This Item

Empirical Formula (Hill Notation):
C10H15N5
CAS Number:
Molecular Weight:
205.26
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$204.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

trapidil

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCN(CC)c1cc(C)nc2ncnn12

InChI

1S/C10H15N5/c1-4-14(5-2)9-6-8(3)13-10-11-7-12-15(9)10/h6-7H,4-5H2,1-3H3

InChI key

GSNOZLZNQMLSKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Trapidil EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Antiplatelet agent; competitive inhibitor of PDGF receptor; phosphodiesterase inhibitor, vasodilator
Trapidil is an antiplatelet agent that acts in part as a phosphodiesterase inhibitor and as a competitive inhibitor of the platelet-derived growth factor (PDGF) receptor. Trapidil, with its vasodilator and NO releasing effect may have some potential to diminish the tissue injury. Trapidil suppresses platelet-derived growth factor (PDGF)-induced vascular smooth muscle cell (VSMC) proliferation by inhibiting Raf-1/extracellular signal-regulated kinase (ERK) via cAMP/protein kinase A (PKA). In addn. to cAMP/PKA activation, trapidil inhibits RhoA/ROCK activation.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yevgeniy V Kalinin et al.
Langmuir : the ACS journal of surfaces and colloids, 25(9), 5391-5397 (2009-03-26)
We study how the microscale topography of a solid surface affects the apparent advancing and receding angles at the contact line of a liquid drop pinned to this surface. Photolithographic methods are used to produce continuous circular polymer rings of
Werner Sziegoleit et al.
Arzneimittel-Forschung, 57(2), 81-86 (2007-04-03)
Since trapidil (CAS 15421-84-8) is able to dilate human hand veins after local intravenous administration, four studies were carried out in healthy male volunteers using the dorsal hand vein compliance technique to test the influence of common systemic single doses
Thorsten Vowinkel et al.
Gastroenterology, 132(3), 955-965 (2007-02-28)
Although the CD40-CD40 ligand (CD40L) signaling pathway has been implicated in the pathogenesis of a variety of diseases, including inflammatory bowel disease, the nature of its contribution to intestinal inflammation remains poorly understood. The aim of this study was to
Neveen A Salem et al.
Renal failure, 32(8), 959-968 (2010-08-21)
Cyclosporine A (CsA) leads to renal and liver injury, production of free radicals and nitric oxide (NO) deficiency. This study investigates the possible protective effects of trapidil and L-arginine against CsA-induced tissue injury. Forty adult male Wistar rats (180 +/-
Salih Somuncu et al.
Pediatric surgery international, 24(3), 315-318 (2007-12-07)
We aimed to detect the protective effect of trapidil in ischemia-reperfusion (IR) injury due to ovarian torsion and detorsion. Thirty-two pubertal New Zealand albino rabbits were used. Adnexal torsion was created by rotating the left adnexa including the tubal and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service