Skip to Content
Merck
All Photos(1)

Key Documents

T1470000

Tinidazole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-[2-(Ethylsulfonyl)ethyl]-2-methyl-5-nitroimidazole

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$313.00

$313.00


Estimated to ship on19 April 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$313.00

About This Item

Empirical Formula (Hill Notation):
C8H13N3O4S
CAS Number:
Molecular Weight:
247.27
Beilstein:
618182
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$313.00


Estimated to ship on19 April 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

tinidazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCS(=O)(=O)CCn1c(C)ncc1[N+]([O-])=O

InChI

1S/C8H13N3O4S/c1-3-16(14,15)5-4-10-7(2)9-6-8(10)11(12)13/h6H,3-5H2,1-2H3

InChI key

HJLSLZFTEKNLFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tinidazole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 2 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gil M Penuliar et al.
Frontiers in microbiology, 6, 354-354 (2015-05-23)
Antimicrobial chemotherapy is critical in the fight against infectious diseases caused by Entamoeba histolytica. Among the drugs available for the treatment of amebiasis, metronidazole (MTZ) is considered the drug of choice. Recently, in vitro studies have described MTZ resistance and
Xian-Shuang Meng et al.
Journal of AOAC International, 97(6), 1538-1545 (2015-01-30)
A sensitive and reliable analytical method based on HPLC/MSIMS has been developed for the simultaneous determination of 15 nitroimidazoles in cosmetics. A diversity of cosmetic samples, including powder, lotion, shampoo, and cream were collected. The samples were ultrasonically extracted with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service