Skip to Content
Merck
All Photos(1)

Key Documents

T1405000

Tiapride hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N-[2-(Diethylamino)ethyl]-2-methoxy-5-(methylsulfonyl)benzamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

30 MG
$254.00

$254.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
30 MG
$254.00

About This Item

Empirical Formula (Hill Notation):
C15H24N2O4S · HCl
CAS Number:
Molecular Weight:
364.89
UNSPSC Code:
41116107
NACRES:
NA.24

$254.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

tiapride

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(C)c1cc(c(cc1)OC)C(=O)NCC[N+H](CC)CC.[Cl-]

InChI

1S/C15H24N2O4S.ClH/c1-5-17(6-2)10-9-16-15(18)13-11-12(22(4,19)20)7-8-14(13)21-3;/h7-8,11H,5-6,9-10H2,1-4H3,(H,16,18);1H

InChI key

OTFDPNXIVHBTKW-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tiapride hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the half life of drug and peak level--Is Diabetes as side effect common or rare

    1 answer
    1. This product is issued by the European Pharmacopeia and it is intended for use as an analytical standard. The half-life is not determined by the supplier Please view the leaflet provided by the European Pharmacopeia: https://crs.edqm.eu/db/4DCGI/leaflet?leaflet=T1405000_2.pdf.

      DrugBank is listing a half-life of 2.9 - 3.6 hours for Tiapride. See the link below for additional information: https://go.drugbank.com/drugs/DB13025

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service