T0918
Terrific Broth, modified
powder
Synonym(s):
TB modified
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
powder
Quality Level
application(s)
food and beverages
suitability
nonselective for Escherichia coli
nonselective for coliforms
Related Categories
Quantity
47.6 g per liter
Preparation Note
1. Suspend 47.6 g and 8 ml glycerol in 1 L of distilled water.
2. Autoclave for 15 minutes at 121°C.
2. Autoclave for 15 minutes at 121°C.
Kit Components Only
Product No.
Description
- Tryptone (pancreatic digest of casein) 12 g/L
- Yeast extract 24 g/L
- K2HPO4 9.4 g/L
- KH2PO4 2.2 g/L
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
AMB Express, 5(1), 70-70 (2015-11-12)
The major platform for high level recombinant protein production is based on genetically modified microorganisms like Escherichia coli (E. coli) due to its short dividing time, ability to use inexpensive substrates and additionally, its genetics is comparatively simple, well characterized
Metabolome analysis reveals a role for glyceraldehyde 3-phosphate dehydrogenase in the inhibition of
Biotechnology for biofuels, 10, 276-276 (2017-12-08)
Here, we analyzed changes in the relative concentration of intracellular metabolites in response to gradual addition of ethanol to growing cultures. For We have demonstrated that a metabolic bottleneck occurs at the GAPDH reaction when the growth of
Science advances, 4(6), eaat1161-eaat1161 (2018-06-22)
Structure and dynamics of living matter rely on design principles fundamentally different from concepts of traditional material science. Specialized intracellular filaments in the cytoskeleton permit living systems to divide, migrate, and grow with a high degree of variability and durability.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service