Skip to Content
Merck
All Photos(1)

Documents

SQC003

Supelco

BNAs in Soil

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

manufacturer/tradename

RTC SQC-003

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

matrix

soil

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    1,2,3,4-Tetrachlorobenzene
    1,2,3,5-Tetrachlorobenzene
    1,2,3-Trichlorobenzene
    1,2,4,5-Tetrachlorobenzene
    1,2,4-Trichlorobenzene
    1,2-Dichlorobenzene
    1,3,5-Trichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    1-Chloronaphthalene
    2,2′-Oxybis(1-Chloropropane)
    2,3,4,6-Tetrachlorophenol
    2,3-Dimethylphenol
    2,4,5-Trichlorophenol
    2,4,6-Trichlorophenol
    2,4-Dichlorophenol
    2,4-Dimethylphenol
    2,4-Dinitrophenol
    2,4-Dinitrotoluene (2,4-DNT)
    2,5-Dimethylphenol
    2,6-Dichlorophenol
    2,6-Dimethylphenol
    2,6-Dinitrotoluene (2,6-DNT)
    2-Amino-1methylbenzene
    2-Chloronaphthalene
    2-Chlorophenol
    2-Methyl-4,6-dinitrophenol
    2-Methylnaphthalene
    2-Methylphenol (o-Cresol)
    2-Nitroaniline
    2-Nitrophenol
    3+4-Methylphenol (m+p-Cresol)
    3,3′-Dichlorobenzidine
    3,4-Dimethylphenol
    3,5-Dimethylphenol
    3-Methylphenol (m-Cresol)
    3-Nitroaniline
    3-Nitrophenol
    4-Bromophenyl phenyl ether
    4-Chloro-3-methylphenol
    4-Chloroaniline
    4-Chlorophenyl phenylether
    4-Methylphenol (p-Cresol)
    4-Nitroaniline
    4-Nitrophenol
    Acenaphthene
    Acenaphthylene
    Acetophenone
    Aniline
    Anthracene
    Atrazine
    Benzaldehyde
    Benzidine
    Benzo(a)anthracene
    Benzo(a)pyrene
    Benzo(b)fluoranthene
    Benzo(b+k)fluoranthene
    Benzo(g,h,i)perylene
    Benzo(k)fluoranthene
    Benzoic acid
    Benzyl alcohol
    Biphenyl
    bis(2-Chloroethoxy)methane
    bis(2-Chloroethyl) ether
    bis(2-Chloroisopropyl) ether
    bis(2-Ethylhexyl) phthalate (DEHP)
    Butyl benzyl phthalate
    Caprolactam
    Carbazole
    Chrysene
    Di-n-butyl phthalate
    Di-n-octyl phthalate
    Dibenzo(a,h)anthracene
    Dibenzofuran
    Diethyl phthalate
    Dimethyl phthalate
    Fluoranthene
    Fluorene
    Hexachlorobenzene
    Hexachlorobutadiene
    Hexachlorocyclopentadiene
    Hexachloroethane
    Indeno(1,2,3-cd) pyrene
    Isophorone
    Maleic anhydride
    n-Nitroso-di-n-propylamine
    n-Nitrosodimethylamine
    n-Nitrosodiphenylamine
    Naphthalene
    Nitrobenzene
    Pentachlorobenzene
    Pentachlorophenol
    Phenanthrene
    Phenol
    Pyrene
    Pyridine
    Total cPAH
    Total PAH
    See All (98)

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Skin Sens. 1

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleksandra Ukalska-Jaruga et al.
Molecules (Basel, Switzerland), 25(3) (2020-02-06)
Pesticides belong to a group of xenobiotics harmful to humans and wildlife, whose fate and activity depends on their susceptibility to degradation. Therefore, the monitoring of their residue level in agricultural soils is very important because it provides very valuable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service