Skip to Content
Merck
All Photos(1)

Documents

SMB01325

Sigma-Aldrich

Albendazole sulfoxide

≥97% (HPLC)

Synonym(s):

Ricobendazole, Albendazole oxide, Albendazole sulfoxide, Methyl [5-(propane-1-sulfinyl)-1H-benzoimidazol-2-yl]-carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N3O3S
CAS Number:
Molecular Weight:
281.33
Beilstein:
677664
MDL number:
UNSPSC Code:
12352119
NACRES:
NA.77

description

Modulation and Signaling

Quality Level

Assay

≥97% (HPLC)

form

solid

color

white to beige

suitability

HPTLC

storage temp.

2-8°C

SMILES string

CCCS(=O)c1ccc2[nH]c(NC(=O)OC)nc2c1

InChI

1S/C12H15N3O3S/c1-3-6-19(17)8-4-5-9-10(7-8)14-11(13-9)15-12(16)18-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)

InChI key

VXTGHWHFYNYFFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Albendazole sulfoxide is a blood-brain barrier main metabolite of anti-parasitic agent albendazole that acts as a antihelminthic. It binds to colchicine binding site of β-tubulin and prevents polymerization. Albendazole sulfoxide exhibits greater affinity to parasitic β-tubulin than to mammalian.

Application

Metabolomics research

Biochem/physiol Actions

Ricobendazole is a key metabolite of albendazole and acts as an anthelmintic. It has been shown to induce apoptosis in human cancer cell line HT-29, possibly by arresting cell cycle at the G2/M phase.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Adrenal gland,spleen,male reproductive organs,Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lidia Radko et al.
Journal of veterinary research, 61(3), 313-319 (2018-07-07)
Albendazole is used to treat endoparasitic diseases in animals and humans. After oral administration, it is quickly oxidised into its pharmacologically active metabolite albendazole sulfoxide and then to sulfone. However, it is not clear which compound is responsible for toxic
Maximo Pettarin et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 155, 105552-105552 (2020-09-17)
The aim of this study was to use a combined in vitro-in silico approach to develop a physiologically based pharmacokinetic model (PBPK) that predicts the bioavailability of albendazole (ABZ), a BCS class II/IV lipophilic weak base, and simulates its main

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service