Skip to Content
Merck
All Photos(1)

Key Documents

SMB00944

Sigma-Aldrich

Dihydrodaidzein

≥95%

Synonym(s):

4′,7-Dihydroxy-isoflavanone; 7,4′-Dihydroxyisoflavanone; 2,3-Dihydro-7-hydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one synthetic

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$284.00

$284.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$284.00

About This Item

Empirical Formula (Hill Notation):
C15H12O4
CAS Number:
Molecular Weight:
256.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

$284.00


Check Cart for Availability

Request a Bulk Order

biological source

synthetic

Quality Level

Assay

≥95%

form

solid

technique(s)

HPLC: suitable

color

faint beige

storage temp.

2-8°C

SMILES string

O1CC(C(=O)c3c1cc(cc3)O)c2ccc(cc2)O

InChI

1S/C15H12O4/c16-10-3-1-9(2-4-10)13-8-19-14-7-11(17)5-6-12(14)15(13)18/h1-7,13,16-17H,8H2

InChI key

JHYXBPPMXZIHKG-UHFFFAOYSA-N

General description

Dihydrodaidzein is an active metabolite of daidzein. It has vasodilatory action on rat isolated aortic rings. It stimulates the estrogen receptor-dependent growth of breast cancer MCF-7 cells at micromolar concentrations. Dihydrodaidzein is produced by the metabolism of daidzein in colonic bacteria and may be further metabolized to various bioactive compounds, including equol .Outside of the human body, Dihydrodaidzein has been detected in several different foods, such as cloves, agars, ohelo berries, greenthread tea, and mexican groundcherries. This could make dihydrodaidzein a potential biomarker for the consumption of these foods.

Application

Metabolomics research

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service