Skip to Content
Merck
All Photos(1)

Key Documents

SBR00029

Sigma-Aldrich

Dansyl labeled polymyxin B Ready Made Solution

for fluorescent microbial imaging, 1.5 mg/mL in H2O

Synonym(s):

DSL-PMB, DSL-PMX, DSL-PxB

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$715.00

$715.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$715.00

About This Item

UNSPSC Code:
51102829
NACRES:
NA.76

$715.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

solution

availability

not available in China

concentration

1.5 mg/mL in H2O

color

colorless to light brown

suitability

suitable for (Fluorescent detection of gram-negative bacteria)
suitable for (Microbiome)
suitable for microbiology

Mode of action

cell membrane

shipped in

ambient

storage temp.

−20°C

General description

Dansyl Polymyxin B (DSL-PMB) is a fluorescent derivative of Polymyxin B. Polymyxin B is a cationic lipopeptide antibiotic that binds specifically to Gram-negative bacteria. Fluorescent antibiotics are obtained by synthetic conjugation of an antibiotic to a fluorophore.
Dansyl labeled Polymyxin B can be utilized for fluorescent microbial imaging in the fields of Cell Biology and Biochemical research.

Application

DSL-PMB may have been used:
  • as a fluorescent probe to study polymyxin mode of action and its pharmacokinetics
  • in research and to develop new active derivatives of Polymyxin against multi-drug resistance Gram-negative infections
  • to measure LPS binding affinity by fluorometric displacement assay
Fluorescent antibiotics can be used for many applications including:
  • Antimicrobial resistance research.
  • Bacterial visualization and imaging.
  • Parent antibiotic mode of action research and new antibiotic discovery.
  • Toxicity studies.
  • Research of bacterial infections and tracking its uptake in vivo.

Biochem/physiol Actions

DSL-PMB′s mode of action is in accordance with Polymyxin B activity. However, MIC assays comparing DSL-PMB, and parent Polymyxin B showed a two to three-fold decrease in bacterial activity. Polymyxin B interacts with anionic lipopolysaccharide (LPS) molecules which are located in Gram-negative bacterial outer membrane. This interaction interferes with the outer membrane′s normal function, causing leakage and enhanced uptake of the antibiotic. The resemblance in properties between Polymyxin B and DSL-PMB allows to fluorescently label Gram-negative bacteria and examine intracellular localization and penetration of Polymyxins in Gram-negative bacteria.

Features and Benefits

  • High-quality antibiotic suitable for multiple research applications
  • Ideal for Cell Biology, Metabolomics, and Biochemical research.

Analysis Note

  • Dansyl Labeled Polymyxin B Ready Made Solution is light sensitive.
  • It is recommended to avoid freeze-thaw cycles of PMB-DSL Ready Made Solution.
  • Dansyl Labeled Polymyxin B Ready Made Solution (1.5 mg/mL) can be diluted 1:50 in PBSX1 (Sigma#D8537) to achieve 30 μg/mL final concentration for staining. The above concentration of DSL-PMB was used for E. coli staining see image.
  • Fluorescence Microscopy application: Dansyl Labeled Polymyxin B Ready Made Solution excitation (Ex) wavelength is 330-340nm resulting in emission (Em) range of 540-600nm (λmax=570nm).

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R A Moore et al.
Antimicrobial agents and chemotherapy, 30(6), 923-926 (1986-12-01)
Pseudomonas cepacia was found to be resistant to the outer membrane-permeabilizing effects of aminoglycoside antibiotics, polymyxin B, and EDTA. Permeabilization of P. cepacia to the fluorescent probe 1-N-phenylnaphthylamine was not achieved at concentrations 100- to 1,000-fold above those required to
R E Hancock
Lancet (London, England), 349(9049), 418-422 (1997-02-08)
The era of the "classical antibiotic" may be over. The emergence of resistance has seen to that. Yet no truly novel class of antibacterial agent has come on the market in the past 30 years. Currently there is great interest
Vincent H Tam et al.
Antimicrobial agents and chemotherapy, 49(9), 3624-3630 (2005-08-30)
Despite limited data, polymyxin B (PB) is increasingly used clinically as the last therapeutic option for multidrug-resistant (MDR) gram-negative bacterial infections. We examined the in vitro pharmacodynamics of PB against four strains of Pseudomonas aeruginosa. Clonal relatedness of the strains
P R Schindler et al.
Antimicrobial agents and chemotherapy, 8(1), 95-104 (1975-07-01)
Though the primary action of the cationic antibiotic polymyxin B is against the membrane of susceptible bacteria, severe morphological changes are detected in the cytoplasm. Using fluorescence microscopy and a mono-N-dansyl-polymyxin B derivative, we could demonstrate aggregations of the antibiotic
Zakuan Z Deris et al.
Bioconjugate chemistry, 25(4), 750-760 (2014-03-19)
The dry antibiotic development pipeline coupled with the emergence of multidrug resistant Gram-negative 'superbugs' has driven the revival of the polymyxin lipopeptide antibiotics. Polymyxin resistance implies a total lack of antibiotics for the treatment of life-threatening infections. The lack of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service