Skip to Content
Merck
All Photos(1)

Key Documents

S2000

General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified

viscosity 4599 mPa.s (25 °C)

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$264.00

$264.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
500 ML
$264.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$264.00


Estimated to ship on01 June 2025


Request a Bulk Order

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

4599 mPa.s(25 °C)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon General Viscosity Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nevena B Ognjenovic et al.
Developmental cell, 55(5), 544-557 (2020-10-30)
Differentiation therapy utilizes our understanding of the hierarchy of cellular systems to pharmacologically induce a shift toward terminal commitment. While this approach has been a paradigm in treating certain hematological malignancies, efforts to translate this success to solid tumors have
Anne K Dettmar et al.
Pediatric nephrology (Berlin, Germany), 33(3), 463-472 (2017-10-17)
Membranous nephropathy (MN) is a common cause of nephrotic syndrome in adults, but is less frequent in children. Antibodies against four antigens leading to MN have been described in children: phospholipase A Twelve children with MN were included in this
Thorsten Wiech et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 32(9), 1320-1328 (2019-04-10)
Renal biopsy is the gold standard for diagnosis of membranous nephropathy. Circulating PLA2R1 antibody found in 75% of patients with membranous nephropathy is very specific for the diagnosis of this disease. Therefore, the question arises whether PLA2R1-antibody-positive patients still need
Christina Zovko et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(58), 13191-13202 (2020-04-15)
A bis(diphenyl)-phosphine functionalized β-diketimine (PNac-H) was synthesized as a flexible ligand for transition metal complexes. The newly designed ligand features symmetrically placed phosphine moieties around a β-diketimine unit, forming a PNNP-type pocket. Due to the hard and soft donor atoms
Jitender Bisht et al.
Lab on a chip, 19(20), 3499-3511 (2019-09-24)
Cell-free Xenopus egg extract is a widely used and biochemically tractable model system that allows recapitulation and elucidation of fundamental cellular processes. Recently, the introduction of microfluidic extract manipulation has enabled compartmentalization of bulk extract and a newfound ability to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service