Skip to Content
Merck
All Photos(1)

Key Documents

S1975000

Sulfafurazole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Sulfisoxazole, 4-Amino-N-(3,4-dimethyl-5-isoxazolyl)benzenesulfonamide, N1-(3,4-Dimethyl-5-isoxazolyl)sulfanilamide, Sulfafurazole

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$204.00

$204.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$204.00

About This Item

Empirical Formula (Hill Notation):
C11H13N3O3S
CAS Number:
Molecular Weight:
267.30
Beilstein:
6737262
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$204.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

sulfafurazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cc1noc(NS(=O)(=O)c2ccc(N)cc2)c1C

InChI

1S/C11H13N3O3S/c1-7-8(2)13-17-11(7)14-18(15,16)10-5-3-9(12)4-6-10/h3-6,14H,12H2,1-2H3

InChI key

NHUHCSRWZMLRLA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulfafurazole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulfafurazole (sulphisoxazole).
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 24, 275-284 (1980-01-01)
[Bacteriology and response to treatment with erythromycin-sulfisoxazole in children with acute otitis media].
R S Rodríguez et al.
Boletin medico del Hospital Infantil de Mexico, 44(12), 728-734 (1987-12-01)
Jang-Gi Choi et al.
PloS one, 9(7), e102697-e102697 (2014-07-23)
Galla rhois is a commonly used traditional medicine for the treatment of pathogenic bacteria in Korea as well as in other parts of Asia. Methyl gallate (MG), a major component of Galla Rhois, exhibits strong antibacterial activity, but its mechanism
Hui-zhen Wu et al.
Analytical and bioanalytical chemistry, 407(13), 3545-3554 (2015-03-04)
An in-line matrix cleanup method was used for the simultaneous extraction of 15 sulfonamides and two metabolites from manure samples. The ultrasound/microwave-assisted extraction (UMAE) combined with solid-liquid-solid dispersive extraction (SLSDE) procedure provides a simple sample preparation approach for the processing
Magda Targa Martins et al.
Talanta, 129, 374-383 (2014-08-17)
In routine laboratory work, screening methods for multiclass analysis can process a large number of samples in a short time. The main challenge is to develop a methodology to detect as many different classes of residues as possible, combined with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service