Skip to Content
Merck
All Photos(2)

Key Documents

R7774

Sigma-Aldrich

Riboflavin 5′-phosphate sodium salt hydrate

meets USP testing specifications

Synonym(s):

Flavin Mononucleotide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$141.00
25 G
$226.00
100 G
$533.00

$141.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
$141.00
25 G
$226.00
100 G
$533.00

About This Item

Empirical Formula (Hill Notation):
C17H20N4NaO9P · xH2O
Molecular Weight:
478.33 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058005
PubChem Substance ID:
NACRES:
NA.21

$141.00


Available to ship on31 March 2025Details


Request a Bulk Order

Agency

USP/NF
meets USP testing specifications

Quality Level

Assay

73.0-79.0% dry basis (BASED ON % RIBOFLAVIN)

form

powder

application(s)

pharmaceutical (small molecule)

storage temp.

−20°C

SMILES string

O.[Na+].Cc1cc2N=C3C(=O)NC(=O)N=C3N(C[C@H](O)[C@H](O)[C@H](O)COP(O)([O-])=O)c2cc1C

InChI

1S/C17H21N4O9P.Na.H2O/c1-7-3-9-10(4-8(7)2)21(15-13(18-9)16(25)20-17(26)19-15)5-11(22)14(24)12(23)6-30-31(27,28)29;;/h3-4,11-12,14,22-24H,5-6H2,1-2H3,(H,20,25,26)(H2,27,28,29);;1H2/q;+1;/p-1/t11-,12+,14-;;/m0../s1

InChI key

BHRVCJBIICJWTH-APQIITSESA-M

General description

Riboflavin 5′-phosphate (flavin mononucleotide (FMN)) is a vitamin B12 derivative, containing isoalloxazine ring system.[1]

Application

Riboflavin 5′-phosphate sodium salt hydrate has been used:
  • as a photosensitizing agent to assess in vitro efficacy of photodynamic therapy (PDT) on fungi[2]
  • to evaluate its in vivo delivery in rat corneas[3]
  • as a component in Vitamin and growth factor stock mix[4]

Biochem/physiol Actions

Riboflavin 5′-phosphate (flavin mononucleotide (FMN)) acts as a coenzyme in intermediary metabolism. Phosphorylation of vitamin B12 by flavokinase yields FMN.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cell isolation and culture
WormBook: The Online Review of C. elegans Biology (2005)
Riboflavin (vitamin B2)
Handbook of vitamins, 255-273 (2013)
Heather Durkee et al.
Lasers in medical science, 35(4), 861-866 (2019-12-25)
To evaluate the in vitro efficacy of rose bengal and riboflavin photodynamic antimicrobial therapy for inhibition the growth of four Pseudomonas aeruginosa (P. aeruginosa) isolates. Four different clinical P. aeruginosa isolates were collected from patients with confirmed keratitis. Each strain
Zhi Li et al.
Biochemical and biophysical research communications, 467(3), 503-508 (2015-10-11)
Desulfotomaculum reducens MI-1 is a Firmicute strain capable of reducing a variety of heavy metal ions and has a great potential in heavy metal bioremediation. We recently identified Dred_2421 as a potential iron reductase through proteomic study of D. reducens. The
Marta Castrillo et al.
Photochemistry and photobiology, 91(6), 1356-1367 (2015-07-29)
Proteins from the cryptochrome/photolyase family utilize UV-A, blue or even red light to achieve such diverse functions as repair of DNA lesions by photolyases and signaling by cryptochromes. DASH-type cryptochromes retained the ability to repair cyclobutane pyrimidine dimers (CPDs) in

Articles

Riboflavin's impact on cell culture systems for manufacturing proteins and monoclonal antibodies discussed.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service