Skip to Content
Merck
All Photos(1)

Key Documents

R0150000

Ranitidine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$221.00

$221.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$221.00

About This Item

Empirical Formula (Hill Notation):
C13H22N4O3S · HCl
CAS Number:
Molecular Weight:
350.86
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$221.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

ranitidine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].CN\C(NCCSCc1ccc(CN(C)C)o1)=C\[N+]([O-])=O

InChI

1S/C13H22N4O3S.ClH/c1-14-13(9-17(18)19)15-6-7-21-10-12-5-4-11(20-12)8-16(2)3;/h4-5,9,14-15H,6-8,10H2,1-3H3;1H/b13-9-;

InChI key

GGWBHVILAJZWKJ-CHHCPSLASA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ranitidine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

H2 histamine receptor antagonist; anti-ulcer agent.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[The development of hemorrhagic vasculitis (Schönlein-Henoch disease) in a patient with a chronic stomach ulcer against a background of Sostril (ranitidine hydrochloride) therapy].
E N Glazunov et al.
Vestnik khirurgii imeni I. I. Grekova, 152(1-2), 102-103 (1994-01-01)
Natalia Alonso et al.
The Biochemical journal, 459(1), 117-126 (2014-01-15)
7TMRs (seven-transmembrane receptors) exist as conformational collections in which different conformations would lead to differential downstream behaviours such as receptor phosphorylation, G-protein activation and receptor internalization. In this context, a ligand may cause differential activation of some, but not all
Question 1: does the use of ranitidine increase the risk of NEC in preterm infants?
Manigandan Chandrasekaran et al.
Archives of disease in childhood, 99(4), 390-392 (2014-03-15)
M B Evans et al.
Journal of pharmaceutical and biomedical analysis, 7(1), 1-22 (1989-01-01)
The selection, development, definition and validation of selective stability-indicating procedures for high-performance liquid chromatographic and thin-layer chromatographic analyses of ranitidine hydrochloride are described. The procedures used in conjunction can be applied to the quality assurance and stability assessments of both
T S Gaginella et al.
Drug intelligence & clinical pharmacy, 17(12), 873-885 (1983-12-01)
Ranitidine is a selective, competitive histamine H2-receptor antagonist recently approved by the Food and Drug Administration for use in the short-term treatment of active duodenal ulcers and gastric hypersecretory conditions. Ranitidine is four to ten times more potent than cimetidine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service