Skip to Content
Merck
All Photos(1)

Key Documents

QC3060

Supelco

Anions - Whole Volume

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

CofA

current certificate can be downloaded

manufacturer/tradename

RTC QCI-3051

concentration

in H2O

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

matrix material

storage temp.

2-8°C

General description

This certified reference material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis. Download your certificate of analysis at: http://www.sigma-aldrich.com.

Preparation Note

The PT sample has been autoclaved for preservation. The sample loses preservation when opened.
The sample must be stored at 4 degrees Celsius.
This sample is a whole volume sample, no dilutions required.

Sample preparation
The sample is a whole volume sample, ready for analysis.
Analyze within 28-days of opening.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Bromide 1 - 10 mg/L
    Chloride 35 - 275 mg/L
    Fluoride 0.4 - 4 mg/L
    Nitrate a N 2 - 25 mg/L
    Nitrate a NO3 1 - 200 mg/L
    Nitrite as N 0.4 - 4 mg/L
    Nitrite a NO2 0 - 100 mg/L
    Orthophosphate as P 0.5 - 5.5 mg/L
    Sulfate 5 - 125 mg/L
    Total nitrate + nitrite as N 2.5 - 25 mg/L
    See All (10)

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service