PHR8705
Dorzolamide Related Compound D
certified reference material, pharmaceutical secondary standard
Synonym(s):
(4S,6S)-4-Amino-6-methyl-5,6-dihydro-4H-thieno[2,3-b]thiopyran-2-sulfonamide 7,7-dioxide hydrochloride, (4S,6S)-4-Amino-6-methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-2-sulfonamide hydrochloride, N-Deethyldorzolamide hydrochloride
Select a Size
$1,350.00
Select a Size
About This Item
$1,350.00
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to USP 1225306
CofA
current certificate can be downloaded
packaging
pkg of 50 mg
application(s)
pharmaceutical
storage temp.
15-25°C
SMILES string
[S]1(=O)(=O)[C@H](C[C@@H](c2c1[s]c(c2)[S](=O)(=O)N)N)C.[Cl-].[H+]
InChI
1S/C8H12N2O4S3.ClH/c1-4-2-6(9)5-3-7(17(10,13)14)15-8(5)16(4,11)12;/h3-4,6H,2,9H2,1H3,(H2,10,13,14);1H/t4-,6-;/m0./s1
InChI key
VZLLBYYSCGNPPR-DPIOYBAHSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Recommended products
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service