Skip to Content
Merck
All Photos(1)

Key Documents

PHR2093

Supelco

Ethylene Oxide and Acetaldehyde Standard Solution

Pharmaceutical Secondary Standard, Certified Reference Material

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$247.00

$247.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$247.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$247.00


Check Cart for Availability

Request a Bulk Order

Quality Level

grade

certified reference material
pharmaceutical secondary standard

CofA

current certificate can be downloaded

packaging

pkg of 5 mL (in amber ampule)

technique(s)

gas chromatography (GC): suitable

application(s)

pharmaceutical small molecule

format

mixture

storage temp.

−20°C

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Components

Ethylene Oxide (0.0010%) and Acetaldehyde (0.0010%) in Polyethylene Glycol 200

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service