Skip to Content
Merck
All Photos(1)

Key Documents

PHR1928

Supelco

Rosuvastatin Calcium

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Rosuvastatin calcium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$462.00

$462.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$462.00

About This Item

Empirical Formula (Hill Notation):
C44H54CaF2N6O12S2
CAS Number:
Molecular Weight:
1001.14
UNSPSC Code:
41116107
NACRES:
NA.24

$462.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001719
traceable to USP 1606015

API family

rosuvastatin

form

powder

CofA

current certificate can be downloaded

packaging

pkg of 1 g

application(s)

pharmaceutical

storage temp.

-10 to -25°C

SMILES string

[Ca+2].Fc3ccc(cc3)c4nc(nc(c4\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])C(C)C)N([S](=O)(=O)C)C.Fc1ccc(cc1)c2nc(nc(c2\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])C(C)C)N([S](=O)(=O)C)C

InChI

1S/2C22H28FN3O6S.Ca/c2*1-13(2)20-18(10-9-16(27)11-17(28)12-19(29)30)21(14-5-7-15(23)8-6-14)25-22(24-20)26(3)33(4,31)32;/h2*5-10,13,16-17,27-28H,11-12H2,1-4H3,(H,29,30);/q;;+2/p-2/b2*10-9+;/t2*16-,17-;/m11./s1

InChI key

LALFOYNTGMUKGG-BGRFNVSISA-L

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Rosuvastatin Calcium, an HMG-CoA reductase inhibitor, belongs to the statins class of pharmaceuticals and is used for the treatment of hypercholesterolemia.[1]

Application

This pharmaceutical secondary standard can also be used as follows:

  • Spectrophotometric estimation of rosuvastatin calcium in its pure form and tablet formulations[1]
  • Determination of rosuvastatin calcium by square-wave voltammetry using a boron-doped diamond electrode in two tablet samples and biological fluid samples of human urine and serum[2]
  • Simultaneous quantification of rosuvastatin and amlodipine by high-performance liquid chromatography (HPLC) in combined pharmaceutical formulations[3]
  • Determination of rosuvastatin calcium and related substances in tablets of rosuvastatin using a reversed-phase high-performance liquid chromatographic (RP-HPLC) method

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2081 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric determination of rosuvastatin calcium in pure form and pharmaceutical formulations by the oxidation using iodine and formation triiodide complex in acetonitrile
Ramadan AA, et al.
International Journal of Pharmacy and Pharmaceutical Sciences, 6, 579-585 (2014)
Simultaneous estimation of rosuvastatin and amlodipine in pharmaceutical formulations using stability indicating HPLC method
Ashfaq M, et al.
Brazilian Journal of Pharmaceutical Sciences , 50, 629-638 (2014)
Square-wave voltammetric determination of rosuvastatin calcium in pharmaceutical and biological fluid samples using a cathodically pretreated boron-doped diamond electrode
Silva TA, et al.
Diamond and Related Materials, 58, 103-109 (2015)

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Please let me know the purity of PHR1928

    1 answer
    1. The purity of this product varies and is listed on the lot specific Certificate of Analysis. Please navigate to the DOCUMENTATION section of the Product Detail Page to access a sample or lot specific Certificate under ‘Certificate of Analysis’:
      https://www.sigmaaldrich.com/product/sial/phr1928#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service