Skip to Content
Merck
All Photos(1)

Key Documents

PHR1816

Supelco

Furosemide Related Compound B

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

4-Chloro-5-sulfamoylanthranilic acid, 4-CHLORO-5- SULFAMOYLANTHRANILIC ACID

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$1,280.00

$1,280.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 MG
$1,280.00

About This Item

Empirical Formula (Hill Notation):
C7H7ClN2O4S
CAS Number:
Molecular Weight:
250.66
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$1,280.00


In StockDetails


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1287030

API family

furosemide

CofA

current certificate can be downloaded

packaging

pkg of 100 mg

mp

267 °C (dec.) (lit.)

application(s)

pharmaceutical

format

neat

storage temp.

2-30°C

SMILES string

Nc1cc(Cl)c(cc1C(O)=O)S(N)(=O)=O

InChI

1S/C7H7ClN2O4S/c8-4-2-5(9)3(7(11)12)1-6(4)15(10,13)14/h1-2H,9H2,(H,11,12)(H2,10,13,14)

InChI key

QQLJBZFXGDHSRU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Furosemide Related Compound B is an impurity of the loop diuretic drug, furosemide.[1] Furosemide is generally used to treat fluid retention in people with heart failure, liver cirrhosis or kidney disease.[2]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Furosemide Related Compound B may be used as a reference standard for the determination of the analyte in bulk drug and pharmaceutical formulations by high performance liquid chromatography.[1][3]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2893 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The analysis of 4-chloro-5-sulfamoylanthranilic acid in the bulk material and pharmaceutical preparations of furosemide by a high-performance liquid chromatographic method
Rapaka RS, et al.
International Journal of Pharmaceutics, 11(3), 237- 247 (1982)
Furosemide
USP42/NF37: United States Pharmacopeia and National Formulary
United States Pharmacopeia/National Formulary, 41(3), 2006-2006 (2018)
Furosemide
StatPearls [Internet] (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service