Skip to Content
Merck
All Photos(1)

Key Documents

PHR1749

Supelco

Trimeprazine Tartrate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Alimemazine hemitartrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$324.00

$324.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$324.00

About This Item

Empirical Formula (Hill Notation):
C18H22N2S · 0.5C4H6O6
CAS Number:
Molecular Weight:
373.49
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$324.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 415
traceable to Ph. Eur. Y0001583
traceable to USP 1689001

API family

trimeprazine

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

S4c5c(cccc5)N(c6c4cccc6)CC(CN(C)C)C.S1c2c(cccc2)N(c3c1cccc3)CC(CN(C)C)C.O[C@H]([C@@H](O)C(=O)O)C(=O)O

InChI

1S/2C18H22N2S.C4H6O6/c2*1-14(12-19(2)3)13-20-15-8-4-6-10-17(15)21-18-11-7-5-9-16(18)20;5-1(3(7)8)2(6)4(9)10/h2*4-11,14H,12-13H2,1-3H3;1-2,5-6H,(H,7,8)(H,9,10)/t;;1-,2-/m..1/s1

InChI key

AJZJIYUOOJLBAU-CEAXSRTFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trimeprazine Tartrate belongs to the group of phenothiazine derivatives,[1] which generally exhibit antipsychotropic, anticholinergic and antihistaminic activities.[2] It is widely used for the treatment of moderate and severe mental and emotional conditions.[1]

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Trimeprazine Tartrate may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using spectrophotometric technique.[1]

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA9455 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flow injection method for the assay of phenothiazine neuroleptics in pharmaceutical preparations using ammonium metavanadate
Sultan MS
Analyst, 116(12), 177-181 (1991)
Flow-injection fluorimetric determination of trimeprazine and trifluoperazine in pharmaceutical preparations
Perez-Ruiz T, et al.
Talanta, 40(9), 1361-1365 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service