Skip to Content
Merck
All Photos(1)

Key Documents

PHR1696

Supelco

Famciclovir

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Famciclovir, 2-(2-(2-Amino-9H-purin-9-yl)ethyl)-1,3-propanediol diacetate ester, BRL 42810

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$449.00

$449.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$449.00

About This Item

Empirical Formula (Hill Notation):
C14H19N5O4
CAS Number:
Molecular Weight:
321.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$449.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1269152

API family

famciclovir

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(=O)OCC(CCn1cnc2cnc(N)nc12)COC(C)=O

InChI

1S/C14H19N5O4/c1-9(20)22-6-11(7-23-10(2)21)3-4-19-8-17-12-5-16-14(15)18-13(12)19/h5,8,11H,3-4,6-7H2,1-2H3,(H2,15,16,18)

InChI key

GGXKWVWZWMLJEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Famciclovir is an antiviral drug, which is predominantly used in the treatment of herpes virus infection. It is an inactive prodrug, which on administration, undergoes rapid biotransformation to penciclovir.[1]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Famciclovir may be used as a reference standard for the determination of the analyte in bulk drug and pharmaceutical formulations by spectrophotometry and high performance liquid chromatography.[2][3][4]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Famciclovir is an Antiviral; Guanine analog used to treat herpesvirus infections.
Famciclovir is an antiretroviral guanosine analog used to treat herpesvirus infections and hepatitis B. Famciclovir is rapidly converted to penciclovir. Viral thymidine kinase phosphorylates penciclovir to a monophosphate form that celular kinases convert in turn to penciclovir triphosphate. Penciclovir triphosphate competitively inhibits viral DNA polymerase and thus viral replication. Prolonged administration can lead to resistance; it is often manifested as selection of pre-existing resistant strains with mutations in the reverse transcriptase domain of the DNA polymerase gene.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3254 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Famciclovir
Perry CM and Wagstaff AJ
Drugs, 50(2), 396-415 (1995)
Development and Validation of liquid chromatographic and UV derivative spectrophotometric methods for the determination of famciclovir in pharmaceutical dosage forms
Srinubabu G, et al.
Chemical & Pharmaceutical Bulletin, 54(6), 819-822 (2006)
Development and validation of LC method for the determination of famciclovir in pharmaceutical formulation using an experimental design
Vishnumulaka S, et al
Journal of Chemistry, 5(1), 58-67 (2008)
Famciclovir
USP42/NF37: United States Pharmacopeia and National Formulary
United States Pharmacopeia/National Formulary, 39(1)(1), 1771-1771 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service