Skip to Content
Merck
All Photos(1)

Key Documents

PHR1659

Supelco

Flucytosine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

5-Fluorocytosine, 4-amino-5-fluoro-2(1H)-pyrimidinone, Flucytosine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$150.00

$150.00


Available to ship on10 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
$150.00

About This Item

Empirical Formula (Hill Notation):
C4H4FN3O
CAS Number:
Molecular Weight:
129.09
Beilstein:
127285
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$150.00


Available to ship on10 April 2025Details


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. F0175000
traceable to USP 1272000

API family

flucytosine

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

298-300 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

NC1=NC(=O)NC=C1F

InChI

1S/C4H4FN3O/c5-2-1-7-4(9)8-3(2)6/h1H,(H3,6,7,8,9)

InChI key

XRECTZIEBJDKEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Flucytosine is an antimycotic drug that can be effectively utilized against severe Candida and Cryptococcus infections.[1]

Application

Flucytosine may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations[1][2][3] by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Nucleoside analog that has antifungal activities. 5-FC is deaminated by cytosine deaminase to product 5-fluorouracil, resulting in RNA miscoding. 5-Fluorocytosine inhibits DNA and RNA synthesis and interferes with ribosomal protein synthesis.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA5760 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stability of flucytosine in an extemporaneously compounded oral liquid.
Wintermeyer SM and Nahata MC
American Journal of Health-System Pharmacy, 53(4), 407-409 (1996)
Analysis of flucytosine dosage forms by derivative UV spectroscopy and liquid chromatography.
Cavrini V, et al.
Journal of Pharmaceutical and Biomedical Analysis, 9(5), 401-407 (1991)
HPLC and GC?MS screening of Chinese proprietary medicine for undeclared therapeutic substances.
Liu SY, et al.
Journal of Pharmaceutical and Biomedical Analysis, 24(5-6), 983-992 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service