Skip to Content
Merck
All Photos(1)

Key Documents

PHR1610

Supelco

Erythromycin Ethylsuccinate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Erythromycin ethyl succinate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$150.00

$150.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$150.00

About This Item

Empirical Formula (Hill Notation):
C43H75NO16
CAS Number:
Molecular Weight:
862.05
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$150.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1245008

API family

erythromycin

CofA

current certificate can be downloaded

packaging

pkg of 250 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)CCC(=O)OC1C(CC(C)OC1OC2C(C)C(OC3CC(C)(OC)C(O)C(C)O3)C(C)C(=O)OC(CC)C(C)(O)C(O)C(C)C(=O)C(C)CC2(C)O)N(C)C

InChI

1S/C43H75NO16/c1-15-29-43(11,52)36(48)24(5)33(47)22(3)20-41(9,51)38(25(6)34(26(7)39(50)57-29)59-32-21-42(10,53-14)37(49)27(8)56-32)60-40-35(28(44(12)13)19-23(4)55-40)58-31(46)18-17-30(45)54-16-2/h22-29,32,34-38,40,48-49,51-52H,15-21H2,1-14H3

InChI key

NSYZCCDSJNWWJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Erythromycin Ethylsuccinate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations[1][2][3] by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA4726 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a stability-indicating high performance liquid chromatography method for assay of erythromycin ethylsuccinate in powder for oral suspension dosage form.
Kamarei F, et al.
Arabian Journal of Chemistry, 7(6), 1079-1085 (2014)
High performance thin layer chromatographic determination of erythromycin in pharmaceutical preparations.
Liawruangrath B and Liawruangrath S
Chromatographia, 54(5-6), 405-408 (2001)
Determination of erythromycin and related substances in enteric-coated tablet formulations by reversed-phase liquid chromatography.
Wardrop J, et al.
Journal of Pharmaceutical Sciences, 89(9), 1097-1105 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service