Skip to Content
Merck
All Photos(1)

Key Documents

PHR1530

Supelco

Acetaminophen Related Compound D

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Acetanilide, N-Phenylacetamide

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
$380.00

$380.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 MG
$380.00

About This Item

Linear Formula:
CH3CONHC6H5
CAS Number:
Molecular Weight:
135.16
Beilstein:
606468
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$380.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1003042

vapor density

4.65 (vs air)

vapor pressure

1 mmHg ( 114 °C)

description

API family: acetaminophen

API family

acetaminophen, paracetamol

CofA

current certificate can be downloaded

autoignition temp.

1004 °F

packaging

pkg of 200 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

304 °C (lit.)

mp

113-115 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CC(=O)Nc1ccccc1

InChI

1S/C8H9NO/c1-7(10)9-8-5-3-2-4-6-8/h2-6H,1H3,(H,9,10)

InChI key

FZERHIULMFGESH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

It is an impurity of the widely used antipyretic and analgesic drug acetaminophen, also known as paracetamol.[1]

Application

This pharmaceutical secondary standard can also be used as follows:

  • Detection and quantification of paracetamol, phenylephrine hydrochloride, and paracetamol impurities in pharmaceutical formulations using four chemometric-based spectrophotometric methods
  • Simultaneous determination of paracetamol and phenylephrine hydrochloride in the presence of paracetamol-related impurities using capillary zone electrophoresis (CZE) in their combined tablet forms[2]

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3648 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

321.8 °F - closed cup

Flash Point(C)

161 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Capillary zone electrophoresis as a quality assessment tool of paracetamol and phenylephrine hydrochloride in presence of paracetamol impurities
FARID J, et al.
Turkish Journal of Chemistry, 46, 217-223 (2022)
Octavian Călinescu et al.
Journal of chromatographic science, 50(4), 335-342 (2012-03-13)
Determination of acetaminophen and its main impurities: 4-nitrophenol, 4'-chloroacetanilide, as well as 4-aminophenol and its degradation products, p-benzoquinone and hydroquinone has been developed and validated by a new high-performance liquid chromatography method. Chromatographic separation has been obtained on a Hypersil
Fabrice Gritti et al.
Journal of chromatography. A, 1355, 179-192 (2014-07-09)
The mass transfer mechanism in four prototype columns (2.1 and 3.0×50mm, 2.1 and 3.0×100mm) packed with 1.9μm fully porous Titan-C18 particles was investigated by using two previously reported home-made protocols. The first one was used to measure the eddy dispersion
Naoki Aizawa et al.
Neurourology and urodynamics, 34(4), 368-374 (2014-02-18)
We measured single-unit mechanosensitive afferent activities (SAAs) during reflexic, rhythmic bladder contractions (RBCs), and examined whether L-arginine, an NO substrate, and mirabegron, a β3-adrenoceptor agonist, and oxybutynin, an antimuscarinic agent, can affect the SAAs in such condition. Twenty-nine female Sprague-Dawley
Hassan A Alhazmi et al.
Journal of pharmaceutical and biomedical analysis, 107, 311-317 (2015-02-02)
In this work, the behavior of several metal ions with different globular proteins was investigated by affinity capillary electrophoresis. Screening was conducted by applying a proper rinsing protocol developed by our group. The use of 0.1M EDTA in the rinsing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service