Skip to Content
Merck
All Photos(1)

Documents

PHR1440

Supelco

Hydroxypropyl Beta Cyclodextrin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352201
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000186
traceable to USP 1329709

API family

cyclodextrin

CofA

current certificate can be downloaded

packaging

pkg of 1 g

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

CC(O)COCC1OC2OC3C(COCC(C)O)OC(OC4C(COCC(C)O)OC(OC5C(COCC(C)O)OC(OC6C(COCC(C)O)OC(OC7C(COCC(C)O)OC(OC8C(COCC(C)O)OC(OC1C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C8OCC(C)O)C(OCC(C)O)C7OCC(C)O)C(OCC(C)O)C6OCC(C)O)C(OCC(C)O)C5OCC(C)O)C(OCC(C)O)C4OCC(C)O)C(OCC(C)O)C3OCC(C)O

InChI

1S/C63H112O42/c1-22(64)8-85-15-29-50-36(71)43(78)57(92-29)100-51-30(16-86-9-23(2)65)94-59(45(80)38(51)73)102-53-32(18-88-11-25(4)67)96-61(47(82)40(53)75)104-55-34(20-90-13-27(6)69)98-63(49(84)42(55)77)105-56-35(21-91-14-28(7)70)97-62(48(83)41(56)76)103-54-33(19-89-12-26(5)68)95-60(46(81)39(54)74)101-52-31(17-87-10-24(3)66)93-58(99-50)44(79)37(52)72/h22-84H,8-21H2,1-7H3/t22?,23?,24?,25?,26?,27?,28?,29-,30-,31?,32?,33?,34?,35?,36?,37-,38?,39-,40+,41+,42+,43?,44+,45?,46+,47+,48+,49+,50+,51+,52-,53-,54-,55-,56-,57+,58-,59+,60-,61-,62-,63-/m1/s1

InChI key

ODLHGICHYURWBS-RYJYQAAZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

Hydroxypropyl Beta Cyclodextrin may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and serum samples by various chromatographic and electroanalytical techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
The solubility of lipophilic drugs increases linearly with the concentration of hydroxypropyl-β-cyclodextrin (HBC) in aqueous solution because of the complex between HBC and the drug. This guest-host type complex is formed between the drug and the non-polar cavity in the HBC that results in enhanced solubility. Solutions may be lyophilized to produce freely soluble powders. Non-toxic in rabbits and mice.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA2748 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Current applications in the analysis of pharmaceuticals by capillary electrophoresis. II
Altria KD, et al.
TrAC, Trends in Analytical Chemistry, 17(4), 214-226 (1998)
Enantiomeric separation of bupropion enantiomers by electrokinetic chromatography: Quantitative analysis in pharmaceutical formulations
Castro-Puyana M, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 875(1), 260-265 (2008)
Separation and determination of norepinephrine, epinephrine and isoprinaline enantiomers by capillary electrophoresis in pharmaceutical formulation and human serum
Wei S, et al.
Journal of Chromatography A, 1098(1-2), 166-171 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service