Skip to Content
Merck
All Photos(1)

Key Documents

PHR1258

Supelco

Chloroquine phosphate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Chloroquine diphosphate salt, N4-(7-Chloro-4-quinolinyl)-N1,N1-dimethyl-1,4-pentanediamine diphosphate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$158.00

$158.00


Available to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
$158.00

About This Item

Empirical Formula (Hill Notation):
C18H26ClN3 · 2H3PO4
CAS Number:
Molecular Weight:
515.86
Beilstein:
4223142
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$158.00


Available to ship on11 April 2025Details


Request a Bulk Order

biological source

synthetic

Quality Level

grade

certified reference material
pharmaceutical secondary standard

Agency

BP
EP
USP
traceable to USP 1118000

vapor pressure

<0.0000001 kPa ( 25 °C)

API family

chloroquine

CofA

current certificate can be downloaded

packaging

pkg of 1 g

storage condition

protect from light

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white to yellow

pH

3.5-4.5 (10% in aqueous solution, pH <6.0)

mp

200 °C (dec.) (lit.)

solubility

water: 50 g/L
benzene: practically insoluble
chloroform: practically insoluble
ethanol: practically insoluble
ether: practically insoluble

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)Nc1ccnc2cc(Cl)ccc12

InChI

1S/C18H26ClN3.2H3O4P/c1-4-22(5-2)12-6-7-14(3)21-17-10-11-20-18-13-15(19)8-9-16(17)18;2*1-5(2,3)4/h8-11,13-14H,4-7,12H2,1-3H3,(H,20,21);2*(H3,1,2,3,4)

InChI key

QKICWELGRMTQCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroquine phosphate is a phosphate salt of chloroquine. It belongs to the class of aminoquinoline drugs utilized as antimalarials and amebicides.[1]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Chloroquine phosphate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by thin-layer chromatography (TLC) and liquid chromatography (LC).[2][3]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB3715 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aminoquinolines against coronavirus disease 2019 (COVID-19): chloroquine or hydroxychloroquine
Sahraei Z, et al.
International Journal of Antimicrobial Agents, 105945(10.1016) (2020)
Determination of chloroquine and its decomposition products in various brands of different dosage forms by liquid chromatography
Abdelrahman AN, et al.
Journal of Pharmaceutical and Biomedical Analysis, 12(2), 205-208 (1994)
Rapid spectrofluorimetric determination of chloroquine phosphate tablets
Nelson O, et al.
International Journal of Drug Development and Research, 2(2), 412-417 (2010)
Alexander Kreuter et al.
Archives of dermatology, 145(3), 244-248 (2009-03-18)
To determine the efficacy of antimalarial drug use in patients with lupus erythematosus tumidus. Retrospective single-center study. Dermatologic clinic at a university hospital. Thirty-six patients with multifocal lupus erythematosus tumidus. Intervention Treatment with either chloroquine phosphate or hydroxychloroquine sulfate. Cutaneous
Claudia G C Cisneros et al.
Acta dermato-venereologica, 90(2), 175-178 (2010-02-20)
Toxic epidermal necrolysis-like lesions have been described in the setting of lupus erythematosus, and have been considered as a specific hyperacute variant of cutaneous lupus erythematosus, with features different from classical drug-related toxic epidermal necrolysis. We report here a series

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service