The correct CAS number for Doxycycline Hyclate is 24390-14-5, which is the material in the vial. The reported assay value vs the USP Reference Standard is expressed as Doxycycline Hyclate because the USP reference standard is Doxycycline Hyclate. It's worth noting that the assay values presented on the Certificate of Analysis are specific to the lot number specified, and these values will differ for each lot number.
PHR1145
Doxycycline hyclate
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Doxycycline hydrochloride hemiethanolate hemihydrate
Select a Size
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to BP 780
traceable to Ph. Eur. D3000000
traceable to USP 1226003
API family
doxycycline
CofA
current certificate can be downloaded
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
-10 to -25°C
SMILES string
Cl[H].Cl[H].[H]O[H].CCO.C[C@@H]1C2[C@H](O)C3[C@H](N(C)C)C(O)=C(C(N)=O)C(=O)[C@@]3(O)C(O)=C2C(=O)c4c(O)cccc14.C[C@@H]5C6[C@H](O)C7[C@H](N(C)C)C(O)=C(C(N)=O)C(=O)[C@@]7(O)C(O)=C6C(=O)c8c(O)cccc58
InChI
1S/2C22H24N2O8.C2H6O.2ClH.H2O/c2*1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)17(27)14-15(24(2)3)18(28)13(21(23)31)20(30)22(14,32)19(12)29;1-2-3;;;/h2*4-7,10,14-15,17,25,27-29,32H,1-3H3,(H2,23,31);3H,2H2,1H3;2*1H;1H2/t2*7-,10?,14?,15-,17-,22-;;;;/m00..../s1
InChI key
HALQELOKLVRWRI-ZVACAFRPSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Doxycycline, the semi-synthetic analog of tetracycline, is a broad-spectrum antibiotic employed in medicine and veterinary applications, is known for its action against a huge range of gram positive and gram negative bacteria. It is prescribed against infections spread by rickettsiae, mycoplasmas and chlamydiae.[1][2]
Application
Biochem/physiol Actions
Activity Spectrum: Effective against a broad spectrum inhibitor of matrix metalloproteinases in vivo.
Analysis Note
Other Notes
Footnote
Recommended products
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.
-
Is Doxycycline Hydrochloride the actual form of the product listed as Doxycycline Hyclate on the Certificate of Analysis (CofA), which states the certified purity as 91.5% Ucrm = +/- 1.1%, l = 2.0?
1 answer-
Helpful?
-
-
Is doxycycline hyclate soluble in DMSO?
1 answer-
According to literature sources, doxycycline hyclate is soluble in DMSO at a concentration of 100 mM.
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service