Skip to Content
Merck
All Photos(1)

Key Documents

PHL89695

Gossypin

phyproof® Reference Substance

Synonym(s):

2-(3,4-Dihydroxyphenyl)-8-(β-D-glucopyranosyloxy)-3,5,7-trihydroxy- 4H-1-benzopyran-4-one, 3,3′,4′,5,7,8-Hexahydroxyflavone 8-glucoside, Gossypetin 8-glucoside

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
$553.00

$553.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 MG
$553.00

About This Item

Empirical Formula (Hill Notation):
C21H20O13
CAS Number:
Molecular Weight:
480.38
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$553.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

SMILES string

OC[C@H]1O[C@@H](Oc2c(O)cc(O)c3C(=O)C(O)=C(Oc23)c4ccc(O)c(O)c4)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H20O13/c22-5-11-13(27)15(29)17(31)21(32-11)34-19-10(26)4-9(25)12-14(28)16(30)18(33-20(12)19)6-1-2-7(23)8(24)3-6/h1-4,11,13,15,17,21-27,29-31H,5H2/t11-,13-,15+,17-,21+/m1/s1

InChI key

SJRXVLUZMMDCNG-KKPQBLLMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Gossypin is an anti-inflammatory pentahydroxyflavone glucoside isolated from Hibiscus vitifolius that is used as an herbal remedy for diabetes, jaundice, and inflammation. Gossypin potently inhibits human cancer cell proliferation, including melanoma and glioma cells. Gossypin directly binds and inhibits BRAFV600E and CDK4 kinases, and also regulates Chk1 and Cdc25C in U251 glioma cells.
Gossypin is an anti-inflammatory pentahydroxyflavone glucoside that directly binds and inhibits BRAFV600E and CDK4 kinases.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service