Skip to Content
Merck
All Photos(1)

Key Documents

PHL89620

Danthrone

phyproof® Reference Substance

Synonym(s):

1,8-Dihydroxyanthraquinone, Chrysazine, Danthron

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$494.00

$494.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$494.00

About This Item

Empirical Formula (Hill Notation):
C14H8O4
CAS Number:
Molecular Weight:
240.21
Beilstein:
2054727
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$494.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

mp

191-193 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

Oc1cccc2C(=O)c3cccc(O)c3C(=O)c12

InChI

1S/C14H8O4/c15-9-5-1-3-7-11(9)14(18)12-8(13(7)17)4-2-6-10(12)16/h1-6,15-16H

InChI key

QBPFLULOKWLNNW-UHFFFAOYSA-N

Gene Information

human ... RXRA(6256)

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Rochowski et al.
International journal of pharmaceutics, 580, 119233-119233 (2020-03-21)
The paper demonstrates the potential of photoacoustics for the identification of the mechanisms underlying drug transport through tissue-mimicking systems. Photoacoustic experiments were performed for a model transdermal delivery system, consisting of drug dithranol (in pharmaceutical form) and dodecanol-collodion (DDC) membrane.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service