Skip to Content
Merck
All Photos(1)

Key Documents

PHL82669

α,β-Thujone

phyproof® Reference Substance

Synonym(s):

(−)-1-Isopropyl-4-methylbicyclo[3.1.0]hexan-3-one

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$471.00

$471.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$471.00

About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$471.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (GC)

manufacturer/tradename

PhytoLab

refractive index

n20/D 1.455 (lit.)

bp

84-86 °C/17 mmHg (lit.)

density

0.925 g/mL at 25 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CC(C)[C@@]12C[C@@H]1[C@@H](C)C(=O)C2.CC(C)[C@@]34C[C@@H]3[C@H](C)C(=O)C4

InChI

1S/2C10H16O/c2*1-6(2)10-4-8(10)7(3)9(11)5-10/h2*6-8H,4-5H2,1-3H3/t7-,8+,10-;7-,8-,10+/m01/s1

InChI key

AKPBIVZAUPVDMO-APEKEWONSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Other Notes

This compound is commonly found in plants of the genus: achillea salvia

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service