Skip to Content
Merck
All Photos(1)

Key Documents

P2800000

Prednisolone acetate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Prednisolone 21-acetate, 1,4-Pregnadiene-11β,17α,21-triol-3,20-dione 21-acetate, 11β,17α,21-Trihydroxy-1,4-pregnadiene-3,20-dione 21-acetate, 21-Acetoxy-1,4-pregnadiene-11β,17α-diol-3,20-dione

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$265.00

$265.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$265.00

About This Item

Empirical Formula (Hill Notation):
C23H30O6
CAS Number:
Molecular Weight:
402.48
Beilstein:
3111798
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$265.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

prednisolone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)COC(C)=O

InChI

1S/C23H30O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h6,8,10,16-18,20,26,28H,4-5,7,9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1

InChI key

LRJOMUJRLNCICJ-JZYPGELDSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Prednisolone acetate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jatin N Ashar et al.
The British journal of ophthalmology, 97(11), 1391-1394 (2013-03-29)
To evaluate a step ladder approach for immunosuppressive regimen for Mooren's ulcer. We retrospectively analysed patients of Mooren's ulcer presenting to a tertiary care centre in south India from 1987 to 2010. Patients were analysed for the age, disease severity
Evan A Olson et al.
Cornea, 31(9), 969-973 (2012-06-09)
To better characterize stromal rejection in the context of deep anterior lamellar keratoplasty (DALK) to improve early diagnosis and proper management. The clinical records of 22 patients undergoing DALK by 2 surgeons between October 2006 and January 2008 were reviewed
Raphael Campagna et al.
European radiology, 23(2), 424-427 (2012-08-07)
To evaluate the occurrence of coring after needle insertion through the rubber stopper of prednisolone acetate vials. Two-hundred vials of prednisolone acetate were randomly distributed to two radiologists. Prednisolone acetate was drawn up through the rubber bung of the vials
Florian Birnbaum et al.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie, 251(1), 95-103 (2012-05-11)
Postoperative astigmatism following penetrating keratoplasty is a major problem after corneal transplantation. The main goal of new trephination techniques such as femtosecond laser or excimer-laser trephination is to improve refractive and visual outcomes. The femtosecond laser technique makes profiled corneal
Susan M Wakil et al.
Canadian journal of ophthalmology. Journal canadien d'ophtalmologie, 47(4), 339-343 (2012-08-14)
The objective of this study was to present a unique case of unilateral facial nerve palsy as an isolated complication of herpes zoster ophthalmicus. An 82-year-old immunocompetent male presented with a 1-week history of painful left scalp lesions. The diagnosis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service