Skip to Content
Merck
All Photos(1)

Documents

P2155017

Plastic additive 17

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Distearyl 3,3′-thiodipropionate, 3,3′-Thiodipropionic acid di-n-octadecyl ester, Dioctadecyl 3,3′-thiodipropionate, Thiodipropionic acid distearyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
S[CH2CH2CO2(CH2)17CH3]2
CAS Number:
Molecular Weight:
683.16
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

EDQM

mp

65-67 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCCCCCCCC

InChI

1S/C42H82O4S/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-37-45-41(43)35-39-47-40-36-42(44)46-38-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-40H2,1-2H3

InChI key

PWWSSIYVTQUJQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Plastic additive 17 EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

458.6 °F

Flash Point(C)

237 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Eelco N Pieke et al.
Analytica chimica acta, 975, 30-41 (2017-05-30)
Risk assessment of exposure to chemicals from food and other sources rely on quantitative information of the occurrence of these chemicals. As screening analysis is increasingly used, a strategy to semi-quantify unknown or untargeted analytes is required. A proof of
Pauline Murat et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 128, 106-118 (2019-03-25)
Container-content interactions are common in the food and pharmaceutical industries. However, these studies are more complicated in the cosmetic industry, and it is necessary to ensure consumer safety. The objective of this work was to develop a strategy for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service