Skip to Content
Merck
All Photos(1)

Key Documents

O3506

Millipore

Oatmeal agar

NutriSelect® Basic, Microbiologically tested, cultivation of fungi, particularly macrospore formation

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$305.00
1 KG
$952.00

$305.00


Available to ship on01 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
$305.00
1 KG
$952.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

$305.00


Available to ship on01 April 2025Details


Request a Bulk Order

Quality Level

sterility

non-sterile

form

powder

composition

Oatmeal 60.0 g/LAgar 12.5 g/L

packaging

pkg of 1 kg
pkg of 250 g

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

final pH

6.0±0.2 (25 °C)

application(s)

clinical testing
food and beverages
food and beverages

suitability

fungi
molds
yeasts

General description

Oatmeal Agar is selective media used for cultivating fungi, particularly for macrospore formation, aiding in their classification and identification. Oatmeal is a source of carbohydrates, amino acids, peptides, vitamins, and minerals for fungal growth. Oatmeal Agar stimulates the organism to produce conidial growth, particularly macroconidia in many filamentous fungal organisms, particularly dermatophytes, as well as chlamydospore formation in yeasts, making it a predominantly suitable medium for the study of fungal morphology and identification. Oatmeal Agar is also a suitable medium for the long-term storage of yeasts and molds because of its low nutrient levels,

Application

Oatmeal Agar is used for cultivating fungi, particularly for macrospore formation from various food, beverages and clinical samples.

Preparation Note

Solve 72.5 g in 1 litre distilled water. Autoclave 15 minutes at 121°C.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colin Charnock
Microorganisms, 9(1) (2021-01-07)
Plastic pollution has become one of the most critical environmental issues, as rapidly increasing production, compounded by persistence of plastic wastes in the environment, are outpacing efforts to keep ecosystems plastic-free. A switch to plastics more amenable to microbial attack

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service