NIST924A
Lithium carbonate
NIST® SRM® 924a
Synonym(s):
Carbolithium, Carbonic acid lithium salt
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
powder
packaging
pkg of 30 g
manufacturer/tradename
NIST®
technique(s)
mass spectrometry (MS): suitable
titration: suitable
mp
618 °C (lit.)
application(s)
clinical
format
neat
SMILES string
[Li+].[Li+].[O-]C([O-])=O
InChI
1S/CH2O3.2Li/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2
InChI key
XGZVUEUWXADBQD-UHFFFAOYSA-L
Gene Information
human ... GSK3A(2931) , GSK3B(2932) , IMPA1(3612)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
SRM 924A_cert
SRM 924A _SDS
Application
Features and Benefits
Other Notes
- Expiration details and storage and use instructions are provided on the NIST certificate
- Notes on biomaterial handling, disposal, and transport documented in MSDS
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service