Skip to Content
Merck
All Photos(1)

Documents

NIST912B

Urea

Synonym(s):

Urea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

description

NIST® SRM® 921b

form

crystals

packaging

pkg of 25 g

manufacturer/tradename

NIST® SRM 912b

technique(s)

Karl-Fischer titration: suitable
NMR: suitable
ion chromatography: suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

Looking for similar products? Visit Product Comparison Guide

General description

Urea Standard Reference Material (SRM) is certified as a neat chemical of known purity. It is a primary standard intended for use during calibration procedures and method validations implemented by clinical laboratories. A unit of SRM 912b consists of 25 g of high-purity crystalline urea.

SRM 912B_cert

SRM 912B_SDS

Application

NIST Urea is intended for use as a primary calibrant for the measurement procedures and validating methods used in clinical pathology laboratories to determine urea blood nitrogen.

Other Notes

  • Expiration details and storage and use instructions are provided on the NIST certificate
  • Notes on biomaterial handling, disposal, and transport documented in MSDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annie Ren et al.
Clinica chimica acta; international journal of clinical chemistry, 542, 117276-117276 (2023-03-05)
We evaluated analytical and Sigma performance for 6 next generation chemistry assays on the Abbott Architect c8000 system. Albumin with bromocresol purple or green, amylase, cholesterol, total protein, and urea nitrogen were analyzed using photometric technology. Analytical performance goals were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service