Skip to Content
Merck
All Photos(1)

Key Documents

NIST8632A

Ultra Fine Test Dust (UFTD)

NIST®SRM®

Synonym(s):

Silica, crystalline quartz

Sign Into View Organizational & Contract Pricing

Select a Size

20 G
$1,950.00

$1,950.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
20 G
$1,950.00

About This Item

Empirical Formula (Hill Notation):
SiO2
CAS Number:
Molecular Weight:
60.08
UNSPSC Code:
41116107
NACRES:
NA.24

$1,950.00


Available to ship on22 April 2025Details


Request a Bulk Order

packaging

pkg of 20 g

manufacturer/tradename

NIST®

application(s)

environmental

format

matrix material

SMILES string

[Si](=O)=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

General description

A unit of RM8632A comprises 20 grams of natural mineral dust exhibiting heterogeneous composition and polydisperse particle size.


SRM 8632A_cert

SRM 8632A_SDS

Application

RM 8632a is intended for use as a secondary test material for validating particle sizing instruments, particularly automatic optical particle counters, when used alongside the established standard method.

Features and Benefits

  • NIST certificate is provided with the reference values for RM 8632a, which represent both the median of the means and the mean of the means for the cumulative particle concentration.
  • The NIST certificate is provided with expiration certificate, use, storage, handling, and maintenance instructions.

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service