Skip to Content
Merck
All Photos(1)

Key Documents

NIST3268

Kudzu (Pueraria montana var. lobata) Extract

NIST®SRM®

Synonym(s):

Kudzu extract

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$1,000.00

$1,000.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$1,000.00

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$1,000.00


Check Cart for Availability

Request a Bulk Order

packaging

pkg of 5 g

manufacturer/tradename

NIST®

format

matrix material

General description

One unit of SRM 3268 contains five packets, each measuring ~1 gram of ground kudzu extract, which can be stored at room temperature.
For more information, please refer to the SDS and COA.
SRM 3268_cert
SRM 3268_SDS

Application
Kudzu Extract Standard Reference Material is intended for evaluating analytical methods used to determine isoflavones and components in ground kudzu extract and similar matrices.
It can also be used for quality assurance while assigning values to in-house reference materials.

Features and Benefits

  • The certified mass fraction values for elements are provided on a dry-mass basis and values aremetrologically traceable to the SI unit of mass fraction, expressed asmilligrams per kilogram (mg/kg).
  • Non-Certified mass fraction values for Isoflavonesin SRM 3268 are provided on a dry-mass basis and values are metrologically traceable to SI unit of massfraction expressed as milligrams per gram.
  • Expiration Certification, Instructions forstorage, and uses are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below.
Arsenic (As), Cadmium (Cd), Lead (Pb), Selenium (Se)

Reference/Informational value is provided for the following analytes.Daidzein (C15H10O4), Daidzin (C21H20O9), Puerarin (C21H20O9)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service