Skip to Content
Merck
All Photos(1)

Documents

NIST2890

Water saturated 1-octanol

NIST® SRM® 2890

Synonym(s):

1-Octanol - water mixture

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 2 mL

manufacturer/tradename

NIST®

technique(s)

Karl-Fischer titration: suitable

format

neat

InChI

1S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3

InChI key

KBPLFHHGFOOTCA-UHFFFAOYSA-N

General description

SRM 2890 is a certified solution of water-saturated 1-octanol. Each unit of SRM 2890 consists of five 5-mL ampoules, with approximately 2 mL of water-saturated 1-octanol.

SRM 2890_cert

SRM 2890_SDS

Application

Water saturated 1-octanol standard reference material (SRM) is intended primarily for use in validating methods and calibrating instruments for the measurement of trace concentration levels of water.

Features and Benefits

  • Available with a certified value of water content that is equally weighted means of the results obtained from:  
(1) Coulometric Karl Fischer measurements.

(2) Volumetric Karl Fischer measurements.

(3) An interlaboratory comparison study involving collaborating laboratories. 

  • Certified values are metrologically traceable to the International System of Units (SI) and SI-derived units of mass. 
  • The NIST certificate is provided with storage, use and handling instructions.

Other Notes

Certified for the analytes listed below.
Water (H2O)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

187.7 °F

Flash Point(C)

86.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Protocols

GC Analysis of Sweet Orange Essential Oil on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service