Skip to Content
Merck
All Photos(1)

Documents

NIST2668

Toxic elements in frozen human urine

NIST® SRM® 2668

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

Quality Level

packaging

pkg of Set (10) (5 ea conc)

manufacturer/tradename

NIST®

application(s)

clinical testing

format

matrix material

General description

Other Notes

Certified for the analytes listed below.
Antimony (Sb), Arsenic (As), Barium (Ba), Beryllium (Be), Cadmium (Cd), Cesium (Cs), Chromium (Cr), Cobalt (Co), Copper (Cu), Lead (Pb), Manganese (Mn), Molybdenum (Mo), Nickel (Ni), Vanadium (V)

Reference/Informational value is provided for the following analytes.
Antimony (Sb), Barium (Ba), Beryllium (Be), Chromium (Cr), Copper (Cu), Manganese (Mn), Molybdenum (Mo), Nickel (Ni), Platinum (Pt), Thallium (Tl), Tin (Sn), Tungsten (W), Uranium (U), Chlorine (Cl), Fluoride (F?), Potassium (K), Sodium (Na), Creatinine (C4H7N3O)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service