Skip to Content
Merck
All Photos(1)

Key Documents

NIST2556

Recycled pellet (autocatalyst)

NIST® SRM® 2556

Sign Into View Organizational & Contract Pricing

Select a Size

70 G
$1,140.00

$1,140.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
70 G
$1,140.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,140.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 70 g

manufacturer/tradename

NIST®

format

matrix material

General description

Other Notes

Certified for the analytes listed below.
Platinum (Pt), Palladium (Pd), Rhodium (Rh),Lead (Pb)

Reference/Informational value is provided for the following analytes. Aluminium (Al), Calcium (Ca), Cerium (Ce), Iron (Fe), Lanthanum (La), Silicon (Si), Barium (Ba), Zinc (Zn), Zirconium (Zr)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Lact. - Repr. 1A - STOT RE 2

Target Organs

Central nervous system,Kidney,Blood

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct determination of trace noble metals (palladium, platinum and rhodium) in automobile catalysts by glow discharge mass spectrometry.
Wayne D M.
Journal of Analytical Atomic Spectrometry, 12(10), 1195-1202 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service