A unit of NIST2135C Ni/Cr thin film depth profile standard (NISTSRM 2135c) consists of nine alternating metal thin-film layers, five layers of pure chromium and four of pure nickel, on a polished silicon (100) substrate. The individual layers have thicknesses that are nominally 57 nm for Cr and 56 nm for Ni. The certified product is 1.0cm x 2.0cm. Please see the NIST document for full details. https://www-s.nist.gov/srmors/certificates/2135c.pdf
NIST2135C
Ni/Cr thin film depth profile standard
NIST SRM 2135c
Select a Size
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
solid
packaging
pkg of each
manufacturer/tradename
NIST®
technique(s)
XRF: suitable
application(s)
environmental
Related Categories
General description
SRM 2135C_cert
SRM 2135C_SDS
Application
Features and Benefits
- The total certified thickness for Chromium (Cr) and Nickel (Ni) single element layer-to-layer uniformity, bilayer uniformity (periodicity) and single layer thickness are expressed in units of mass/area and determined by XRF and ICP using a gravimetrically calibrated reference.
- Expiration details and instructions for use, storage and handling are provided on the NIST certificate.
Other Notes
Nickel (Ni), Chromium (Cr)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the size and shape of the NIST2135C Ni/Cr thin film depth profile standard?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service